Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,O-Bis(trimethylsilyl)trifluoroacetamide
N,O-Bis(trimethylsilyl)trifluoroacetamide
N,O-Bis(trimethylsilyl)trifluoroacetamide
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide
Supplier:
Restek
Certificates
Expand image
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide
Supplier:
Restek
About this item
CAS Number:
25561-30-2
:
Formula:
C₈H₁₈F₃NOSi₂
MDL Number:
MFCD00008269
:
257.4 g/mol
UN:
1993
ADR:
3,II
:
GHS02
:
GHS05