Orders LinkContactUsLinkComponent
Ferrule removal kit
Ferrule removal kit
Ferrule removal kit
Catalog # REST20146
Supplier:   Restek
Ferrule removal kit
Catalog # REST20146
Supplier:  Restek
Ferrule removal kit
Ferrule removal kit
Ferrule removal kit
Price
98,89 €
Log in for availability
1 * 1 items
Product Details & Documents
The tapered tools in this kit have teeth designed to grip and remove fused silica adapter ferrules that have become stuck in the fitting detail.
Delivery information: Each kit contains two tools: one for removing 0,8 mm adapter ferrules and one for removing 1,6 mm adapter ferrules.
Specifications
  • Description:
    Ferrule removal kit