Tubing reamer
Catalog # REST20134
Supplier: Restek
Specifications
- Description:Tubing reamer
- Pk:1
Specifications
About this item
The tubing reamer removes burrs from stainless steel tubing.
- For 3,1 to 6,35 mm tubing
- Nonslip safety design
The tubing reamer removes burrs from stainless steel tubing.