FID/injector cleaning kit
Catalog # REST20120
Supplier: Restek
Specifications
- Description:FID/injector cleaning kit
- Pk:1
Specifications
About this item
This FID/injector cleaning kit can be used wih any injection ports.
Delivery: Kit includes nylon tube brushes of sizes (¹/₈", ³/₁₆" and ¹/₄"), pipe cleaner, stainless steel tube brushes of sizes (³/₈", ³/₁₆" and ¹/₄") and surface brush, stainless steel jet reamers and emery cloth.