Orders LinkContactUsLinkComponent
Cut resistant gloves, HyFlex® 11-541
Supplier:   Ansell
Cut resistant gloves, HyFlex® 11-541
Supplier:  Ansell
Certificates
Cut resistant gloves, HyFlex® 11-541
6 Options Available Below
Product Details & Documents
These seamless liners with INTERCEPT™ technology yarn feature a nitrile foam coating on palm and fingertips.
  • Form-fitting ultralight weight glove
  • ANSELL GRIP™ Technology minimises the force required to grip oily or wet tools or materials
  • INTERCEPT™ Technology yarn has been designed to exceed industry standards
  • Best-in-class cut protection and increased comfort with dexterity
  • FORTIX™ Technology applies a thin, resilient and breathable nitrile foam coating, which extends gloves' working life and improves comfort in abrasive applications
Ideal for the assembly of small sharp parts, for the handling of small sharp metal parts, light duty stamping and maintenance.
 : EN 388:2016 (4X21D), EN 407 (X1XXXX); Cat. III.
Specifications for Product Family
Specifications
  • Catalog No:
  • ANSE11-541/6
  • ANSE11-541/7
  • ANSE11-541/8
  • ANSE11-541/9
  • ANSE11-541/10
  • ANSE11-541/11
  • Model:
  • 11-541
  • 11-541
  • 11-541
  • 11-541
  • 11-541
  • 11-541
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • PE
  • PE
  • PE
  • PE
  • PE
  • PE
  • Finish:
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Gauge:
  • 18
  • 18
  • 18
  • 18
  • 18
  • 18
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • EN 388:2016 (4X21D), EN 407 (X1XXXX)
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Length
    Packed
    Availability
    Price
  • 6
    Grey
    200 - 250 mm
    12 pairs/bag
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 417,00 €
    417,00 €
    1 * 12 PAIR
       417,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ANSE11-541/6
    Model
    11-541
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    PE
    Finish
    Palm and fingertips coated
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • 7
    Grey
    200 - 250 mm
    12 pairs/bag
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 417,00 €
    417,00 €
    1 * 12 PAIR
       417,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ANSE11-541/7
    Model
    11-541
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    PE
    Finish
    Palm and fingertips coated
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • 8
    Grey
    200 - 250 mm
    12 pairs/bag
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 417,00 €
    417,00 €
    1 * 12 PAIR
       417,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ANSE11-541/8
    Model
    11-541
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    PE
    Finish
    Palm and fingertips coated
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • 9
    Grey
    200 - 250 mm
    12 pairs/bag
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 417,00 €
    417,00 €
    1 * 12 PAIR
       417,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ANSE11-541/9
    Model
    11-541
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    PE
    Finish
    Palm and fingertips coated
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • 10
    Grey
    200 - 250 mm
    12 pairs/bag
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 417,00 €
    417,00 €
    1 * 12 PAIR
       417,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ANSE11-541/10
    Model
    11-541
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    PE
    Finish
    Palm and fingertips coated
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X21D), EN 407 (X1XXXX)
  • 11
    Grey
    200 - 250 mm
    12 pairs/bag
    Log in for availability
    Log in for availability
    1 * 12 PAIR
    1 * 12 PAIR - 417,00 €
    417,00 €
    1 * 12 PAIR
       417,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ANSE11-541/11
    Model
    11-541
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    PE
    Finish
    Palm and fingertips coated
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (4X21D), EN 407 (X1XXXX)