Orders LinkContactUsLinkComponent
Preassembled vial kits
Preassembled vial kits
Supplier:   Restek
Preassembled vial kits
Supplier:  Restek
Certificates
Preassembled vial kits
Preassembled vial kits
8 Options Available Below
Product Details & Documents
Vial kits with 8 mm/425 screw-thread, unmarked vials, septa and caps (caps are attached to vials).
Product Family Options
Product Information
  • Capacity
    Colour
    Cap included
    Cap type
    Cap Size
    Cap colour
    Septum
    Thickness
    Availability
    Price
  • 2,00 ml
    Clear
    Yes
    Screw
    8-425
    Black
    Red PTFE/silicone
    1,14 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 72,00 €
    72,00 €
    1 * 100 items
       72,00 €
  • 2,00 ml
    Clear
    Yes
    Screw
    8-425
    Black
    Red PTFE/silicone
    1,14 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 637,00 €
    637,00 €
    1 * 1.000 items
       637,00 €
  • 2,00 ml
    Clear
    Yes
    Screw
    8-425
    Black
    Red PTFE/silicone
    1,65 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 72,00 €
    72,00 €
    1 * 100 items
       72,00 €
  • 2,00 ml
    Clear
    Yes
    Screw
    8-425
    Black
    Red PTFE/silicone
    1,65 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 618,00 €
    618,00 €
    1 * 1.000 items
       618,00 €
  • 2,00 ml
    Amber
    Yes
    Screw
    8-425
    Black
    Red PTFE/silicone
    1,14 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 78,00 €
    78,00 €
    1 * 100 items
       78,00 €
  • 2,00 ml
    Amber
    Yes
    Screw
    8-425
    Black
    Red PTFE/silicone
    1,14 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 690,00 €
    690,00 €
    1 * 1.000 items
       690,00 €
  • 2,00 ml
    Amber
    Yes
    Screw
    8-425
    Black
    Red PTFE/silicone
    1,65 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 78,00 €
    78,00 €
    1 * 100 items
       78,00 €
  • 2,00 ml
    Amber
    Yes
    Screw
    8-425
    Black
    Red PTFE/silicone
    1,65 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 671,00 €
    671,00 €
    1 * 1.000 items
       671,00 €