Sapphire scribe
Catalog # REST20182
Supplier: Restek
Specifications
- Description:Sapphire scribe
- Pk:1
Specifications
About this item
This sapphire scribe cuts fused silica tubing.
- Produces a clean, square cut
This sapphire scribe cuts fused silica tubing.