Order Entry
Export
ContactUsLinkComponent
IL-33 (human) ELISA kit
IL-33 (human) ELISA kit
Catalog # ENZOADI900201
Supplier:  Enzo Life Sciences
CAS Number:  
IL-33 (human) ELISA kit
Catalog # ENZOADI900201
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-33
  • Description:
    IL-33 ELISA (human) kit, 96 wells
  • Seq:
    02

Specifications

About this item

IL-33 human ELISA Kit is a colorimetric immunometric immunoassay kit for the quantitative determination of human IL-33 in cell lysates, culture supernatants, serum, plasma and synovial fluid samples.

  • Sensitive - reliably measure as little as 1,7 pg/ml of IL-33
  • Low dynamic range provides sensitive measurements
  • Specific - low cross reactivity to other interleukins
  • Non-radioactive - enzymatic assay format is safe and reliable
  • Easy to use - liquid colour coded reagents reduce errors
Delivery: Kit contains microtiter plate, conjugate, antibody, assay buffer 13, wash buffer concentrate, standard, TMB substrate, stop solution 2, xtraction reagent concentrate.