ContactUsLinkComponent
Bulk adsorbents, Ottawa sand
Bulk adsorbents, Ottawa sand
Catalog # REST26137
Supplier:  Restek
CAS Number:  
undefined
Bulk adsorbents, Ottawa sand
Catalog # REST26137
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Ottawa sand bulk adsorbent, 20/30 mesh
Specifications
About this item
Ottawa sand is listed in several U.S. EPA methods as the specified medium for matrix spike and laboratory control blanks.
Packaging: Packaged in convenient 5 kg buckets.