Cleaning brushes for extractor cells
Cell cleaning brushes, Nylon, Black, Wire
Catalog # REST23999
Supplier: Restek
Jump to:
Product Details & Documents
Firm nylon bristle brushes for easy cell cleaning and removal of solid samples.
Range of sizes to fit all extraction cells for ASE® 100, 150, 200, 300, and 350.
Specifications
- Bristle colour:Black
- Bristle material:Nylon
- Handle material:Wire
- Description:Cell cleaning brushes