ContactUsLinkComponent
Bulk adsorbents, sodium sulfate
  REST26204
 :  Restek
 :  
undefined
Bulk adsorbents, sodium sulfate
  REST26204
 :  Restek
 :  
  • Pk:
    5 kg
  • Description:
    Sodium sulfate bulk adsorbent, 10-60 mesh
Anhydrous sodium sulphate is the most common drying agent used to remove moisture from sample extracts. Activate by heating to 400 °C for four hours before use.
 : Packaged in recloseable 5 kg buckets.