Orders LinkContactUsLinkComponent
Columns, MXT®-5
Supplier:   Restek
Columns, MXT®-5
Supplier:  Restek
Certificates
Columns, MXT®-5
Columns, MXT®-5
43 Options Available Below
Product Details & Documents
These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.
  • Equivalent to USP G27, G36 phases
  • 4,5 inch standard coil diameter
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • REST70205
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,10 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 573,00 €
    573,00 €
    1 * 1 items
       573,00 €
  • REST70208
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,10 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 939,00 €
    939,00 €
    1 * 1 items
       939,00 €
  • REST71821
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,20 µm
    0,18 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 707,00 €
    707,00 €
    1 * 1 items
       707,00 €
  • REST71822
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,20 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 936,00 €
    936,00 €
    1 * 1 items
       936,00 €
  • REST71823
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,20 µm
    0,18 mm
    40 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 408,00 €
    1 408,00 €
    1 * 1 items
       1 408,00 €
  • REST70220
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 573,00 €
    573,00 €
    1 * 1 items
       573,00 €
  • REST70221
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,28 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 583,00 €
    583,00 €
    1 * 1 items
       583,00 €
  • REST70222
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 599,00 €
    599,00 €
    1 * 1 items
       599,00 €
  • REST70223
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 929,00 €
    929,00 €
    1 * 1 items
       929,00 €
  • REST70224
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,28 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 947,00 €
    947,00 €
    1 * 1 items
       947,00 €
  • REST70225
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 035,00 €
    1 035,00 €
    1 * 1 items
       1 035,00 €
  • REST70226
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 530,00 €
    1 530,00 €
    1 * 1 items
       1 530,00 €
  • REST70227
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,28 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 595,00 €
    1 595,00 €
    1 * 1 items
       1 595,00 €
  • REST70228
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,25 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 638,00 €
    1 638,00 €
    1 * 1 items
       1 638,00 €
  • REST71824
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,40 µm
    0,18 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 707,00 €
    707,00 €
    1 * 1 items
       707,00 €
  • REST71825
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,40 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 936,00 €
    936,00 €
    1 * 1 items
       936,00 €
  • REST70235
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,50 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 568,00 €
    568,00 €
    1 * 1 items
       568,00 €
  • REST70236
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,50 µm
    0,28 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • REST70237
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 599,00 €
    599,00 €
    1 * 1 items
       599,00 €
  • REST70238
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,50 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 929,00 €
    929,00 €
    1 * 1 items
       929,00 €
  • REST70239
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,50 µm
    0,28 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 947,00 €
    947,00 €
    1 * 1 items
       947,00 €
  • REST70240
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 035,00 €
    1 035,00 €
    1 * 1 items
       1 035,00 €
  • REST70241
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    0,50 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • REST70250
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,00 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 568,00 €
    568,00 €
    1 * 1 items
       568,00 €
  • REST70251
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,00 µm
    0,28 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 583,00 €
    583,00 €
    1 * 1 items
       583,00 €
  • 3,5" coil
    Capillary
    MXT®-5
    1,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 599,00 €
    599,00 €
    1 * 1 items
       599,00 €
  • REST70252
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 599,00 €
    599,00 €
    1 * 1 items
       599,00 €
  • REST70253
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 929,00 €
    929,00 €
    1 * 1 items
       929,00 €
  • REST70254
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,00 µm
    0,28 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 947,00 €
    947,00 €
    1 * 1 items
       947,00 €
  • 3,5" coil
    Capillary
    MXT®-5
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 035,00 €
    1 035,00 €
    1 * 1 items
       1 035,00 €
  • REST70255
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 035,00 €
    1 035,00 €
    1 * 1 items
       1 035,00 €
  • REST70257
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,00 µm
    0,28 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 595,00 €
    1 595,00 €
    1 * 1 items
       1 595,00 €
  • REST70258
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 638,00 €
    1 638,00 €
    1 * 1 items
       1 638,00 €
  • REST70267
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,50 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 599,00 €
    599,00 €
    1 * 1 items
       599,00 €
  • REST70270
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    1,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 035,00 €
    1 035,00 €
    1 * 1 items
       1 035,00 €
  • REST70281
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    3,00 µm
    0,28 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • REST70282
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    3,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 599,00 €
    599,00 €
    1 * 1 items
       599,00 €
  • REST70284
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    3,00 µm
    0,28 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 947,00 €
    947,00 €
    1 * 1 items
       947,00 €
  • REST70285
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 035,00 €
    1 035,00 €
    1 * 1 items
       1 035,00 €
  • REST70288
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    3,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 638,00 €
    1 638,00 €
    1 * 1 items
       1 638,00 €
  • REST70277
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    5,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 616,00 €
    616,00 €
    1 * 1 items
       616,00 €
  • REST70279
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    5,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 043,00 €
    1 043,00 €
    1 * 1 items
       1 043,00 €
  • REST70283
    4,5" standard coil
    4,5" standard coil
    Capillary
    MXT®-5
    5,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 638,00 €
    1 638,00 €
    1 * 1 items
       1 638,00 €