Anti-Synj1 Mouse Monoclonal Antibody [clone: 5H1]
Catalog # ABNOMAB14170
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Synj1 Recombinant
- Antigen symbol:Synj1
- Clonality:Monoclonal
- Clone:5H1
- Host:Mouse
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:85238
- Amino acid number:Q62910
- Storage buffer:In HEPES, 0.15 M NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
- Immunogen:Recombinant protein corresponding to C-terminus of rat Synj1.
- Purification:Purified
- Size:100 µL
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant rat Synj1.
Type: Primary
Antigen: Synj1
Clonality: Monoclonal
Clone: 5H1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rat