Anti-Rxrb Mouse Monoclonal Antibody [clone: 147]
Catalog # ABNOMAB14153
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:Rxrb
- Clonality:Monoclonal
- Clone:147
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Form:Liquid
- Gene ID:361801
- Amino acid number:P49743
- Storage buffer:In HEPES, 0.15 M NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
- Immunogen:A hinge peptide corresponding to human, mouse and rat Rxrb.
- Purification:Purified
- Size:100 µL
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against rat Rxrb.
Type: Primary
Antigen: Rxrb
Clonality: Monoclonal
Clone: 147
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat