Order Entry
Export
ContactUsLinkComponent
 
Anti-RARB Mouse Monoclonal Antibody [clone: 336]
Catalog # ABNOMAB14152
Supplier:  Abnova
Anti-RARB Mouse Monoclonal Antibody [clone: 336]
Catalog # ABNOMAB14152
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Retinoic Acid Receptor Beta
  • Antigen symbol:
    RARB
  • Clonality:
    Monoclonal
  • Clone:
    336
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    5915
  • Amino acid number:
    P10826
  • Storage buffer:
    In HEPES, 0.15 M NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
  • Immunogen:
    A synthetic peptide corresponding to N-terminus of human RARB.
  • Purification:
    Purified
  • Size:
    100 µL
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of human RARB.

Type: Primary
Antigen: RARB
Clonality: Monoclonal
Clone: 336
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human