Anti-KRT18 Mouse Monoclonal Antibody [clone: KRT18/834]
ABNOMAB14336
: Abnova
- Antibody type:Primary
- Antigen name:KRT18 Recombinant
- Antigen symbol:KRT18
- Clonality:Monoclonal
- Clone:KRT18/834
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:3875
- Amino acid number:P05783
- Storage buffer:In 10 mM PBS.
- Immunogen:Recombinant protein corresponding to full length human KRT18.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human KRT18.
Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: KRT18/834
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human