Anti-KRT18 Mouse Monoclonal Antibody [clone: DE-K18]
ABNOMAB14333
: Abnova
- Antibody type:Primary
- Antigen name:Keratin 18
- Antigen symbol:KRT18
- Clonality:Monoclonal
- Clone:DE-K18
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3875
- Amino acid number:P05783
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Cytoskeletal preparation extracted from the human vulvar squamous carcinoma cell line A431.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Mouse monoclonal antibody raised against native human KRT18.
Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: DE-K18
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human