Anti-IGKC Mouse Monoclonal Antibody [clone: Kap-56]
Catalog # ABNOMAB14289
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:IGKC
- Clonality:Monoclonal
- Clone:Kap-56
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3514
- Amino acid number:P01834
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Human lymphocytes stimulated.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human Ig kappa light chain.
Type: Primary
Antigen: IGKC
Clonality: Monoclonal
Clone: Kap-56
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human