Anti-IGHM Mouse Monoclonal Antibody [clone: ICO-30]
Catalog # ABNOMAB14288
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:IGHM
- Clonality:Monoclonal
- Clone:ICO-30
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3507
- Amino acid number:P01871
- Storage buffer:In 10 mM PBS.
- Immunogen:Human IgM heavy chain.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against native human IgM.
Type: Primary
Antigen: IGHM
Clonality: Monoclonal
Clone: ICO-30
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human