Order Entry
Export
ContactUsLinkComponent
 
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: 19-26.1(MB-26.1)]
  ABNOMAB14281
 :  Abnova
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: 19-26.1(MB-26.1)]
  ABNOMAB14281
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    HLA-DRA
  • Clonality:
    Monoclonal
  • Clone:
    19-26.1(MB-26.1)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    3122
  • Amino acid number:
    P01903
  • Storage buffer:
    In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
  • Immunogen:
    Raji cells.
  • Purification:
    Protein A/G purification
  • Size:
    100 µG
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against native human HLA-DRA.

Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: 19-26.1(MB-26.1)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human