Order Entry
Export
ContactUsLinkComponent
Anti-CGB Mouse Monoclonal Antibody (CF488A) [clone: HCGb/54 HCGb/459]
Anti-CGB Mouse Monoclonal Antibody (CF488A) [clone: HCGb/54 HCGb/459]
Supplier:  Biotium
Certificates
Anti-CGB Mouse Monoclonal Antibody (CF488A) [clone: HCGb/54 HCGb/459]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC881224-100
  • BNC881224-500
  • Antigen symbol:
  • CGB
  • CGB
  • Antigen name:
  • Glycoprotein hormones, beta polypeptide
  • Glycoprotein hormones, beta polypeptide
  • Antigen synonyms:
  • Choriogonadotropin Subunit beta|CGB3|CG-beta|CGB8|CGB5|hCGB|CGB7
  • Choriogonadotropin Subunit beta|CGB3|CG-beta|CGB8|CGB5|hCGB|CGB7
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone HCGb/54 HCGb/459
  • Clone HCGb/54 HCGb/459
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 172944
  • 172944
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications