Anti-CD63 Mouse Monoclonal Antibody (PE) [clone: MX-49.129.5]
Catalog # ABNOMAB14212
Supplier: Abnova
Specifications
- Antibody type:Secondary
- Antigen symbol:CD63
- Clonality:Monoclonal
- Clone:MX-49.129.5
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:967
- Amino acid number:P08962
- Storage buffer:In PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Native purified human CD63.
- Purification:Protein G purification
- Size:500 µL
- Pk:500 µl
Specifications
About this item
Mouse monoclonal antibody raised against native human CD63.
Type: Secondary
Antigen: CD63
Clonality: Monoclonal
Clone: MX-49.129.5
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse