Anti-SIRT6 Rabbit Polyclonal Antibody
Catalog # ABGEAP14024B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Sirtuin 6
- Antigen symbol:SIRT6
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:SIR2-like protein 6|Mono-ADP-ribosyltransferase sirtuin-6|Silent information regulator 2-like protein 6.
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SIRT6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human