Order Entry
Export
ContactUsLinkComponent
Anti-FCGR2B Rabbit Polyclonal Antibody
Anti-FCGR2B Rabbit Polyclonal Antibody
  ABGEAP13987B
 :  Abgent
Anti-FCGR2B Rabbit Polyclonal Antibody
  ABGEAP13987B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen symbol:
    FCGR2B
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: FCGR2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human