Anti-PCP2 Rabbit Polyclonal Antibody
Catalog # ABGEAP6356B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:PCP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Epitope:C-Terminal
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse