Order Entry
Export
ContactUsLinkComponent
Anti-CD66 molecule Mouse Monoclonal Antibody [clone: C66/195 C66/261 C66/1009 C66/1030]
Anti-CD66 molecule Mouse Monoclonal Antibody [clone: C66/195 C66/261 C66/1009 C66/1030]
Catalog # BNUM1139-50
Supplier:  Biotium
Anti-CD66 molecule Mouse Monoclonal Antibody [clone: C66/195 C66/261 C66/1009 C66/1030]
Catalog # BNUM1139-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD66
  • Antigen symbol:
    CD66 molecule
  • Clonality:
    Monoclonal
  • Clone:
    Clone C66/195 C66/261 C66/1009 C66/1030
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Gene ID:
    709196
  • Antigen synonyms:
    CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci of gastric carcinoma and in distinguishing pulmonary adenocarcinomas (60-70% are CEA ) from pleural mesotheliomas (rarely or weakly CEA ). Anti-CEA positivity is seen in adenocarcinomas from the lung, colon, stomach, esophagus, pancreas, gallbadder, urachus, salivary gland, ovary, and endocervix.

  • Purified
  • BSA-free

Type: Primary
Antigen: CD66 molecule
Clonality: Monoclonal
Clone: C66/195 C66/261 C66/1009 C66/1030
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human