Anti-AKT3 Rabbit Polyclonal Antibody
Catalog # ABGEAP7030C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:V-Akt Murine Thymoma Viral Oncogene homolog 3
- Antigen symbol:AKT3
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Epitope:Center
- Antigen synonyms:PKB-GAMMA|PKBG|RAC-PK-gamma|STK-2|RAC-gamma|PRKBG|MPPH2|MPPH
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: