About this item
β2 microglobulin is a 12 kDa protein with a pI of 5.6. Serum β2 microglobulin levels are a reflection of cell turnover. Levels rise with fever, inflammation, and infection. Increased serum levels are also seen in B-cell malignancies and in renal failure and may indicate a worse prognosis for patients with early-stage Hodgkin's lymphoma. In urine, increased levels are seen in proximal renal tubular disease as well as renal transplant rejection. β2 microglobulin levels can rise either because its rate of synthesis has increased (e.g. in AIDS, malignant monoclonal plasma cell dyscrasia, solid tumours and autoimmune disease) or because of impaired renal filtration (e.g. due to renal insufficiency, graft rejection or nephrotoxicity induced by post-transplantation immunosuppressive therapy).
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: B2M/1118
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa1 kappa
Reactivity: Human, Non-human primates
Specifications
- Catalog No:
- BNC041118-100
- BNC041118-500
- Antigen symbol:
- B2M
- B2M
- Antigen name:
- Beta-2-Microglobulin
- Beta-2-Microglobulin
- Antigen synonyms:
- Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
- Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Clone B2M/1118
- Clone B2M/1118
- Reactivity:
- Primates, Human
- Primates, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 534255
- 534255
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide