Order Entry
Export
ContactUsLinkComponent
Anti-HLA-DRA Mouse Monoclonal Antibody (Biotin) [clone: 19-26.1]
Anti-HLA-DRA Mouse Monoclonal Antibody (Biotin) [clone: 19-26.1]
Supplier:  Biotium
Certificates
Anti-HLA-DRA Mouse Monoclonal Antibody (Biotin) [clone: 19-26.1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB1082-500
  • BNCB1082-100
  • Antigen symbol:
  • MHC2
  • MHC2
  • Antigen name:
  • MHC II
  • MHC II
  • Antigen synonyms:
  • HLA-DRA
  • HLA-DRA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone 19-26.1
  • Clone 19-26.1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 520048
  • 520048
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications