Anti-KRT76 Mouse Monoclonal Antibody (Biotin) [clone: KRTH/1076]
Supplier: Biotium
Certificates
About this item
This MAb recognizes basic (Type II or HMW) cytokeratins, which include 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8). Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50 , 48, 46, 45, and 40 kDa. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Type: Primary
Antigen: KRT76
Clonality: Monoclonal
Clone: KRTH/1076
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Specifications
- Catalog No:
- BNCB1076-100
- BNCB1076-500
- Antigen symbol:
- KRT76
- KRT76
- Antigen name:
- Cytokeratin HMW
- Cytokeratin HMW
- Antigen synonyms:
- Keratin|Type-II Keratin Kb9|Cytokeratin-2P|KRT2P|Cytokeratin 2|HUMCYT2A|K2P|CK-2P|Type II Cytoskeletal 2 Oral|K76|Keratin 2p|Keratin-76|KRT2B
- Keratin|Type-II Keratin Kb9|Cytokeratin-2P|KRT2P|Cytokeratin 2|HUMCYT2A|K2P|CK-2P|Type II Cytoskeletal 2 Oral|K76|Keratin 2p|Keratin-76|KRT2B
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone KRTH/1076
- Clone KRTH/1076
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Mouse
- Mouse
- Gene ID:
- 654392
- 654392
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide