Order Entry
Export
ContactUsLinkComponent
Anti-Human Papillomavirus 16 Mouse Monoclonal Antibody (CF405S) [clone: HPV16/1058]
Anti-Human Papillomavirus 16 Mouse Monoclonal Antibody (CF405S) [clone: HPV16/1058]
Supplier:  Biotium
Certificates
Anti-Human Papillomavirus 16 Mouse Monoclonal Antibody (CF405S) [clone: HPV16/1058]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC041058-500
  • BNC041058-100
  • Antigen name:
  • Human Papillomavirus 16
  • Human Papillomavirus 16
  • Antigen synonyms:
  • HPV-16 capsid|HPV16 L1|HPV-16|HPV16 major capsid protein L1|Human papillomavirus type 16 major capsid protein L1|Human papillomavirus type 16 L1
  • HPV-16 capsid|HPV16 L1|HPV-16|HPV16 major capsid protein L1|Human papillomavirus type 16 major capsid protein L1|Human papillomavirus type 16 L1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone HPV16/1058
  • Clone HPV16/1058
  • Reactivity:
  • Human papilloma virus (HPV)
  • Human papilloma virus (HPV)
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications