Order Entry
Export
ContactUsLinkComponent
Anti-CD48 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 5-4.8]
Anti-CD48 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 5-4.8]
Catalog # BNUM1057-50
Supplier:  Biotium
Anti-CD48 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 5-4.8]
Catalog # BNUM1057-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD48
  • Clonality:
    Monoclonal
  • Clone:
    Clone 5-4.8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Gene ID:
    243564
  • Antigen synonyms:
    BCM1 surface antigen|B cell membrane protein|SLAM family member 2 (SLAMF2)|BLAST1|Signaling lymphocytic activation molecule 2|TCT.1|B-cell activation marker
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Reacts with human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of the markers for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 5-4.8
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human