ACTH (same as Corticotropin) is a 39 amino acid active peptide produced by the anterior pituitary. This MAb is specific to Synacthen (aa1-24 of ACTH); does not react with CLIP (aa17-39 of ACTH). POMC (pro-opiomelanocortin or corticotropin-lipotropin) is a 267 amino acid polypeptide hormone precursor that goes through extensive, tissue-specific posttranslational processing by convertases. POMC is cleaved into ten hormone chains named NPP, ACTH, alpha-MSH (Melanocyte Stimulating Hormone), beta-MSH, gamma-MSH, CLIP (corticotropin-like intermediary peptide), Lipotropin-beta, Lipotropin-gamma, beta-endorphin and Met-enkephalin. ACTH is also produced by cells of immune system (T-cells, B-cells, and macrophages) in response to stimuli associated with stress. Anti-ACTH is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with ACTH-producing cells (corticotrophs).It also may react with other tumors (e.g. some small cell carcinomas of the lung) causing paraneoplastic syndromes by secreting ACTH.
Type: Primary
Antigen: ACTH
Clonality: Monoclonal
Clone: AH26 57
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- BNC681025-100
- BNC681025-500
- Antigen symbol:
- ACTH
- ACTH
- Antigen name:
- Adrenocorticotropic hormone
- Adrenocorticotropic hormone
- Antigen synonyms:
- alpha or beta or gamma Melanocyte Stimulating Hormone (MSH) or Melanotropin|beta-Endorphin|POC|beta or gamma Lipotropin (LPH)|CLIP|POMC|Adrenocorticotropin|Met Enkephalin
- alpha or beta or gamma Melanocyte Stimulating Hormone (MSH) or Melanotropin|beta-Endorphin|POC|beta or gamma Lipotropin (LPH)|CLIP|POMC|Adrenocorticotropin|Met Enkephalin
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone AH26 57
- Clone AH26 57
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 1897
- 1897
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide