Anti-RBFOX3 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-77686B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:RNA Binding Protein, Fox-1 Homolog 3
- Antigen symbol:RBFOX3
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Gene ID:146713
- Antigen synonyms:FOX-3|FOX3|NEUN|HRNBP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: RBFOX3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat