Anti-CD84 Mouse Monoclonal Antibody (Biotin) [clone: 152-1D5]
: Biotium
Recognizes a protein of 74 kDa, identified as CD84 (Workshop V; Code A085, C057). It is expressed on mature B cells and on B-cell lines, including pre-B-cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly N-glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation.
Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: 152-1D5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
- Catalog No:
- BNCB0985-100
- BNCB0985-500
- Antigen symbol:
- CD84
- CD84
- Antigen synonyms:
- LY9B|Signaling lymphocytic activation molecule 5 (SLAMF5)|hCD84|Cell surface antigen MAX.3|SLAM family member 5|Hly9-beta|Leucocyte differentiation antigen CD84
- LY9B|Signaling lymphocytic activation molecule 5 (SLAMF5)|hCD84|Cell surface antigen MAX.3|SLAM family member 5|Hly9-beta|Leucocyte differentiation antigen CD84
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone 152-1D5
- Clone 152-1D5
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 398093
- 398093
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide