Anti-KRT5 & KRT8 Mouse Monoclonal Antibody (CF488A) [clone: C-50]
Supplier: Biotium
Certificates
About this item
This antibody reacts with keratin 5 (58 kDa) and keratin 8 (52.5 kDa). Simple epithelia express cytokeratin 8 in combination with 18. On the other hand, basal cells of stratified epithelia express cytokeratin 5 paired with 14. This antibody therefore, reacts with a wide range of epithelia and their carcinomas.
Type: Primary
Antigen: KRT5 & KRT8
Clonality: Monoclonal
Clone: C-50
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Dog, Ferret, Hamster, Human, Mouse, Pig, Rat, Sheep
Specifications
- Catalog No:
- BNC880948-100
- BNC880948-500
- Antigen symbol:
- KRT5 & KRT8
- KRT5 & KRT8
- Antigen name:
- Cytokeratin 5/8
- Cytokeratin 5/8
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone C-50
- Clone C-50
- Reactivity:
- Hamster, Pig, Sheep, Ferret, Human, Bovine, Dog, Rat, Mouse
- Hamster, Pig, Sheep, Ferret, Human, Bovine, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 433845 (CK5); 533782 , 708445 (CK8)
- 433845 (CK5); 533782 , 708445 (CK8)
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide