Order Entry
Export
ContactUsLinkComponent
 
Anti-DPP4 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 202-36]
Catalog # BNUM0888-50
Supplier:  Biotium
Anti-DPP4 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 202-36]
Catalog # BNUM0888-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dipeptidylpeptidase 4
  • Antigen symbol:
    DPP4
  • Clonality:
    Monoclonal
  • Clone:
    Clone 202-36
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Rat
  • Gene ID:
    368912
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Recognizes a glycoprotein of 110 kDa, identified as CD26 (Workshop VI; Code: N-L039). It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross-linking of CD26 using this antibody dramatically enhances the anti-CD3-induced IL-2 production. In Western blotting, this MAb reacts with only glycosylated CD26, but not with the deglycosylated form. It does not prevent ADA binding to CD26.

Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: 202-36
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rat