Order Entry
Export
ContactUsLinkComponent
Anti-DPP4 Mouse Monoclonal Antibody (CF568) [clone: 202-36]
Anti-DPP4 Mouse Monoclonal Antibody (CF568) [clone: 202-36]
Supplier:  Biotium
Certificates
Anti-DPP4 Mouse Monoclonal Antibody (CF568) [clone: 202-36]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC680888-100
  • BNC680888-500
  • Antigen symbol:
  • DPP4
  • DPP4
  • Antigen name:
  • Dipeptidylpeptidase 4
  • Dipeptidylpeptidase 4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone 202-36
  • Clone 202-36
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 368912
  • 368912
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications