About this item
This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the kappa light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant.
Type: Primary
Antigen: IGKC
Clonality: Monoclonal
Clone: Kap-56
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC940859-100
- BNC940859-500
- Antigen symbol:
- IGKC
- IGKC
- Antigen name:
- Kappa Light Chain
- Kappa Light Chain
- Antigen synonyms:
- Ig Kappa Chain C Region|IGKC|HCAK1|Immunoglobulin KM
- Ig Kappa Chain C Region|IGKC|HCAK1|Immunoglobulin KM
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone Kap-56
- Clone Kap-56
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 449609
- 449609
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide