Anti-TGFA Mouse Monoclonal Antibody (CF568) [clone: P/T1]
Supplier: Biotium
Certificates
About this item
This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: P/T1
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rabbit, Zebrafish
Specifications
- Catalog No:
- BNC680787-100
- BNC680787-500
- Antigen symbol:
- TGFA
- TGFA
- Antigen name:
- TGFalpha
- TGFalpha
- Antigen synonyms:
- TGFA|Waved 1|ETGF|TFGA|TGF Type 1|Wa1|EGF-like TGF
- TGFA|Waved 1|ETGF|TFGA|TGF Type 1|Wa1|EGF-like TGF
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone P/T1
- Clone P/T1
- Reactivity:
- Zebrafish, Rabbit, Human
- Zebrafish, Rabbit, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 170009
- 170009
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide