Anti-IGHM Mouse Monoclonal Antibody (CF568) [clone: IM260 IM373 ICO-30]
Supplier: Biotium
Certificates
About this item
Recognizes a protein of 75 kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Type: Primary
Antigen: IGHM
Clonality: Monoclonal
Clone: IM260 IM373 ICO-30
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Specifications
- Catalog No:
- BNC680762-100
- BNC680762-500
- Antigen symbol:
- IGHM
- IGHM
- Antigen name:
- IgM Immunoglobulin
- IgM Immunoglobulin
- Antigen synonyms:
- Ig Mu Chain C Region|Constant Region of Heavy Chain of IgM|AGM1|IGHM
- Ig Mu Chain C Region|Constant Region of Heavy Chain of IgM|AGM1|IGHM
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone IM260 IM373 ICO-30
- Clone IM260 IM373 ICO-30
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 510635
- 510635
- Isotype:
- IgGs
- IgGs
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide