Anti-PRLR Mouse Monoclonal Antibody (CF647) [clone: B6.2 PRLR742]
Supplier: Biotium
Certificates
About this item
This antibody recognizes a protein of 70 kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.
Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: B6.2 PRLR742
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Specifications
- Catalog No:
- BNC470744-500
- BNC470744-100
- Antigen symbol:
- PRLR
- PRLR
- Antigen name:
- Prolactin Receptor
- Prolactin Receptor
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone B6.2 PRLR742
- Clone B6.2 PRLR742
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 368587
- 368587
- Isotype:
- IgGs
- IgGs
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide