Anti-HTR3A Rabbit Polyclonal Antibody
BOSSBS-12051R
: Bioss
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine (serotonin) receptor 3A, ionotropic
- Antigen symbol:HTR3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:3359
- Antigen synonyms:5-HT3A|HTR3|5HT3R|5-HT-3|5-HT3R
- Storage buffer:Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at −20 °C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4 °C
- Immunogen:20-70/478
- Purification:Purified by Protein A
- Size:100 μl
- Pk:100 µl
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses in neurons. It is a cation-specific, but otherwise relatively nonselective, ion channel.
Recommended Dilutions: Western Blot: 1:100 to 1000; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.
Type: Primary
Antigen: HTR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat