Order Entry
Export
ContactUsLinkComponent
Anti-Keratins Mouse Monoclonal Antibody (CF647) [clone: KRT/457]
Anti-Keratins Mouse Monoclonal Antibody (CF647) [clone: KRT/457]
Supplier:  Biotium
Certificates
Anti-Keratins Mouse Monoclonal Antibody (CF647) [clone: KRT/457]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470457-100
  • BNC470457-500
  • Antigen symbol:
  • Multiple
  • Multiple
  • Antigen name:
  • Cytokeratin multi
  • Cytokeratin multi
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone KRT/457
  • Clone KRT/457
  • Reactivity:
  • Frog, Pig, Monkey, Goat, Human, Bovine, Guinea pig, Rat, Mouse
  • Frog, Pig, Monkey, Goat, Human, Bovine, Guinea pig, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications