Anti-Keratins Mouse Monoclonal Antibody (Biotin) [clone: KRT/457]
Supplier: Biotium
Certificates
About this item
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, including 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8); 56.5 kDa (CK10); 53 kDa (CK13) and 45 kDa (CK18). This is a broad-spectrum antibody, which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Type: Primary
Antigen: Keratins
Clonality: Monoclonal
Clone: KRT/457
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Frog, Goat, Guinea Pig, Human, Monkey, Mouse, Pig, Rat
Specifications
- Catalog No:
- BNCB0457-100
- BNCB0457-500
- Antigen symbol:
- Multiple
- Multiple
- Antigen name:
- Cytokeratin multi
- Cytokeratin multi
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone KRT/457
- Clone KRT/457
- Reactivity:
- Frog, Pig, Monkey, Goat, Human, Bovine, Guinea pig, Rat, Mouse
- Frog, Pig, Monkey, Goat, Human, Bovine, Guinea pig, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide