This MAb reacts with a membrane-associated protein present in normal and malignant neuroendocrine tissues including small cell lung cancer (SCLC). It stains neural and a variable number of endocrine tissues and in the lung it reacts preferentially with SCLC and carcinoids. Its epitope is destroyed during formalin fixation. This antibody was categorized during the First International Workshop on Small Cell Lung Cancer Antigens held in London in April 1987. There are two major types of Lung Carcinoma: non-small cell, which accounts for 80% of all cases; and small cell, which accounts for roughly 20% of all lung cancers reported. The lung continues to be a customary place for cancer migration from tumors elsewhere in the body. Treatment depends on the specific cell type of the cancer, level of progression and status of the individual patient.
- Purified
Type: Primary
Antigen: Small Cell Lung Cancer
Clonality: Monoclonal
Clone: MOC-52
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
- Catalog No:
- BNUB0329-100
- BNUB0329-500
- Antigen name:
- Small Cell Lung Cancer
- Small Cell Lung Cancer
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone MOC-52
- Clone MOC-52
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide