Order Entry
Export
ContactUsLinkComponent
Anti-KRT76 Mouse Monoclonal Antibody (CF405S) [clone: AE-3]
Anti-KRT76 Mouse Monoclonal Antibody (CF405S) [clone: AE-3]
Supplier:  Biotium
Certificates
Anti-KRT76 Mouse Monoclonal Antibody (CF405S) [clone: AE-3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040257-100
  • BNC040257-500
  • Antigen symbol:
  • KRT76
  • KRT76
  • Antigen name:
  • Cytokeratin HMW
  • Cytokeratin HMW
  • Antigen synonyms:
  • Keratin|Type-II Keratin Kb9|KRT2P|Cytokeratin-2P (CK-2P|Keratin 2p (K2P)|type II Cytoskeletal 2 oral|HUMCYT2A|K76|Keratin-76|KRT2B
  • Keratin|Type-II Keratin Kb9|KRT2P|Cytokeratin-2P (CK-2P|Keratin 2p (K2P)|type II Cytoskeletal 2 oral|HUMCYT2A|K76|Keratin-76|KRT2B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone AE-3
  • Clone AE-3
  • Reactivity:
  • Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 654392
  • 654392
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications