Order Entry
Export
ContactUsLinkComponent
 
Anti-KRT77 Mouse Monoclonal Antibody (CF640R) [clone: AE-1]
 :  Biotium
 
Anti-KRT77 Mouse Monoclonal Antibody (CF640R) [clone: AE-1]
 :  Biotium

 

  • Catalog No:
  • BNC400253-100
  • BNC400253-500
  • Antigen symbol:
  • KRT77
  • KRT77
  • Antigen name:
  • Cytokeratin LMW
  • Cytokeratin LMW
  • Antigen synonyms:
  • K77|Keratin|KRT1B|Type-II Keratin Kb39|Keratin-77|type II cytoskeletal 1b|K1B|CK-1B|Keratin 1B|Cytokeratin-1B
  • K77|Keratin|KRT1B|Type-II Keratin Kb39|Keratin-77|type II cytoskeletal 1b|K1B|CK-1B|Keratin 1B|Cytokeratin-1B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone AE-1
  • Clone AE-1
  • Reactivity:
  • Turtle, Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Turtle, Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 334989
  • 334989
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide