Anti-THY1 Mouse Monoclonal Antibody (Purified) [clone: F15-42-1]
Supplier: Biotium
Certificates
About this item
CD90 (Thy-1) is an 18-35 kDa GPI-anchored plasma membrane glycoprotein expressed in many cell types, such as in hematopoietic cells and neurons, connective tissues, various fibroblast and stromal cell lines, tumor endothelial cell lines and others. It is involved in T-cell activation, cellular adhesion, proliferation and migration, neurite outgrowth, wound healing, apoptosis, and fibrosis. CD90 participates in multiple signaling cascades and its effects are tissue- and cell type-specific. It often functions as an important regulator of cell-cell and cell-matrix interactions.
Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey
Specifications
- Catalog No:
- BNUB0194-500
- BNUB0194-100
- Antigen symbol:
- THY1
- THY1
- Antigen name:
- Thy1 Cell Surface Antigen
- Thy1 Cell Surface Antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Clone F15-42-1
- Clone F15-42-1
- Reactivity:
- Monkey, Human
- Monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 644697
- 644697
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide