Order Entry
Export
ContactUsLinkComponent
 
Anti-CD59 Mouse Monoclonal Antibody (Purified) [clone: 193-27]
 :  Biotium
 
Anti-CD59 Mouse Monoclonal Antibody (Purified) [clone: 193-27]
 :  Biotium

 

  • Catalog No:
  • BNUB0182-100
  • BNUB0182-500
  • Antigen symbol:
  • CD59
  • CD59
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone 193-27
  • Clone 193-27
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 278573; 709466 , 710641
  • 278573; 709466 , 710641
  • Isotype:
  • IgM kappa
  • IgM kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide